United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
9 >
EW-86515-01
Drager PAC 3500 Single-Gas Monitors, O2
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86515-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17102-07
Drager PAC Multi-Gas Calibration
$135.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Gas Detectors
>
Single Gas Detection
>
Drager Pac 3500/5500 Single-Gas Monitors
Share
|
Drager PAC 3500 Single-Gas Monitors, O2
EW-86515-01
$207.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Three alarms functionsaudible, visual, and vibrationmake alarm conditions clear
The monitor features a robust, rubber-coated weatherproof polymer housing rated to IP65 that will stand up to the typical abuse in an industrial environment. Easily operated using just two buttons. Its small size makes it comfortable and easy to wear all day on a belt, pocket, or lapel. A loud audible alarm, two bright visual alarms, and strong vibrating alarm alert you to hazardous concentrations of gas. Built-in event logger stores up to 60 alarm activations or other events.The Pac 3500 is a maintenance-free detector with a two-year sensor life.
Approval:
UL and cUL classified intrinsically safe in Class I, Div. 1, Groups A, B, C, D, E, F, G T4; ATEX; CE
What's included:
Pocket/belt clip and batteries.
Specifications
Product Type
Single Gas Monitors/Detectors
Resolution
0.1%
Gas
Oxygen
Range
0 to 25%
Sensor life
2 years
Alarm
90 dB (audible), visual, vibration
Alarm set points
Low: 19.5%
High: 23%
Dimensions
3.25 in x 2.5 in x 1 in
(8.4 x 6.4 x 2.5 cm)
Manufacturer number
4543959
Power
Nonreplaceable lithium batteries
CE Compliance
Yes
Brand
Drager
Calibrations
Qty
/
Price
Availability
Drager PAC Multi-Gas Calibration
EW-17102-07
at $135.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Armed_for_unseen_hazards
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Isolating Genomic DNA from Whole Blood
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title