log in or register
Drum Pump, 5 Strokes/gallon, 316 SS And PTFE
EW-07079-90
$99.00 USD/EACH
  • Ideal for most organic phase applications
  • Suitable for pumping chlorinated or petroleum-based solvents
  • 316 SS/PTFE® parts offer excellent chemical resistance for detergents, acids, corrosives and other liquids compatible with 316 SS and PTFE.
  • T-shaped handle for comfortable grip
  • Larger outlet spout provides greater flowrate
Steel hand-operated drum pumps feature a convenient 1⁄2" OD spout.To safely pump petroleum-based solvents, use grounding wire (order separately see "Accessories").
What's Included:
vented bung adapter. Order optional adapters for 5-gallon containers (see "Accessories").
Accessories
Qty/Price
Availability
at $113.00 USD/EACH
Usually ships in 13 days.
at $14.00 USD/EACH
Usually ships in 8 days.
at $11.00 USD/EACH
In stock.
at $44.00 USD/EACH
Usually ships in 12 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title