United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
6 >
EW-04318-20
Heavy-duty mixer 04318-15 with C-clamp mount shown as sample only. See individual product details.
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04318-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-03
Innocal NIST Calibration for Mixer
$115.00 USD/EACH/EACH
Usually ships in 13 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Industrial Mixers
>
Drum and Barrel Mixers
>
Drum and Barrel Heavy-Duty Electric Motor Mixer
Share
|
Drum/barrel heavy-duty mixer with drum lip mounting; 1/2 hp, 115/230 VAC
EW-04318-20
$947.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Mixer
Impeller size
3.75 in (folding)
Motor (hp)
0.5
Mounting type
drum-lip
Shaft length
33.125 in
Max torque (in-oz)
288
Accessories
Qty
/
Price
Availability
Lightnin 304 SS marine impeller, 4.0" dia x 5/8" bore dia
EW-50509-10
at
$84.00 USD/EACH
In stock.
Add to Cart
Folding impeller, 3-3/4" dia (max) x 5/8" bore dia
EW-50509-20
at
$155.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Mixer
EW-17050-03
at $115.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title