United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Drum barrel tube mixer 115 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04316-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Industrial Mixers
>
Drum and Barrel Mixers
>
Drum and Barrel Top-To-Bottom Tube Mixers
>
Drum and Barrel Top-to-Bottom Electric Motor Tube Mixers
Share
|
Drum/barrel tube mixer, 115 VAC
EW-04316-20
$1,694.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Top-to-bottom movement provides thorough mixing without creating a vortex
These efficient tube mixers allow you to thoroughly mix the contents of 55-gallon (200 liter) drums without removing the drum lid. The 40" (1016 mm) long mixing tube fits into standard 2" (51 mm) bungs and generates a powerful top-to-bottom flow.
A shaft-mounted turbine inside the mixing tube creates a steady suction effect at four upper intake ports. Mixers draw approximately 13 to 14 gallons (49 to 53 liters) of liquid (750 cp max) per minute through the 2" (51 mm) OD mixing tube. The forcefully expelled liquid resuspends and dissolves settled particles. Choose either an electric or air-drive motor mixer.
Electric motor mixers
have 1/4-hp motors that maintain a 3450-rpm mixing speed and operate on 115 VAC, 60 Hz. Mixers feature an on/off switch on the motor for your convenience. Select either a totally enclosed, fan-cooled (TEFC) motor or CSA-approved explosion-proof motor (rated for Class 1, Division 1, Group D conditions).
What's Included:
40"L x 2" OD (1016 mm L x 51 mm OD) mixing tube with PTFE resin bearing at the top and base; 316 SS shaft; turbine and sleeve, and 10-ft (3-m), 3-wire cord with plug (electric mixers only).
Specifications
Product Type
Mixer
Max speed (rpm)
3450
Motor (hp)
1/4
Motor type
TEFC
Power (VAC)
115
Power (Hz)
60
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title