log in or register
Dry Block Calibrator, Tecal 140F, 120V, Model FDB140FS
EW-16102-05
$4,470.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Housed in a rugged metal case for field use
  • High accuracy and stability for certifying temperature sensors and hydraulic thermostats
  • Fast settling time for rapid testing
  • Fast heat up and cool down times
  • Large interchangeable insert format for flexibility of use
  • RS-232 computer interface for data and program transfer, or real time control
  • Improved uniformity and control stability of the temperature zone
F Series
Following on from over 20 years of experience with Dry Block Calibrators, the new F models have been introduced to offer a compact, economical and easy-to-use high performance range of Dry Block temperature calibrators.
  • Display resolution up to 0.01°C (0.1°F)
  • Built rugged for field work
  • Simple to use and operate
  • Custom inserts for tapered and/or nonstandard sensor types available
  • Large multi-well inserts to calibrate several sensors at one time
  • RS-232 interface
    H Series
  • 4 line by 20 character LCD and 8 button keypad for entering programs and viewing data
  • The ability to create, use and store programs
    Product Type Block Calibrators
    Description 140F Series
    Resolution 0.01°C or 0.1°F
    Max temperature 140°C
    Temp accuracy 0.3°C
    Temp uniformity 0.2°C
    Power 120 V, 50/60 Hz, 400 W
  • Accessories
    Qty/Price
    Availability
    at $182.00 USD/EACH
    Usually ships in 10 days.
    at $499.00 USD/EACH
    Usually ships in 9 days.
    7002590:Rs232 Cable
    EW-16102-03
    at $25.50 USD/EACH
    Call 1-888-358-4705 for availability.
    at $182.00 USD/EACH
    In stock.
    2 X1/4inAND2 X3/8in
    EW-16104-26
    at $182.00 USD/EACH
    Usually ships in 7 days.
    2 1/4in/2 X 1/2in
    EW-16104-27
    at $182.00 USD/EACH
    Usually ships in 7 days.
    at $182.00 USD/EACH
    Call 1-888-358-4705 for availability.
    7032538:Blank Insert
    EW-16104-29
    at $182.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $406.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $507.00 USD/EACH
    Call 1-888-358-4705 for availability.
    1X 9/16AND1 X 1/4in
    EW-16104-39
    at $262.00 USD/EACH
    Usually ships in 7 days.
    1 X 5/8in 1 X 1/4in
    EW-16104-40
    at $262.00 USD/EACH
    Usually ships in 9 days.
    at $262.00 USD/EACH
    Usually ships in 7 days.
    1 X 3/4inAND1X1/4in
    EW-16104-42
    at $262.00 USD/EACH
    Call 1-888-358-4705 for availability.
    140F And Tecal 650F
    EW-16104-48
    at $219.00 USD/EACH
    Call 1-888-358-4705 for availability.
    425f
    EW-16104-49
    at $171.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Calibrations
    Qty/Price
    Availability
    at $250.00 USD/EACH
    Usually ships in 8 days.
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title