United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
4
Representative photo only
Dry Glass Bead Sterilizer 5 D x 1 3 4 dia sterilizing well 220 VAC 50 Hz 175 Watts
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10779-15
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Sterilizers
>
Dry Glass Bead Sterilizers
Share
|
Dry Glass Bead Sterilizer, 5" D x 1-3/4" dia sterilizing well; 220 VAC, 50 Hz, 175 Watts
EW-10779-15
$788.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Effectively sterilize small, solid metal, and glass implements in seconds
Heated glass beads destroy all microbes and spores
Low power consumption allows for all day operation
Safe to useno gases, fumes, open flames or harsh liquids
Sterilize solid metal and glass implements in less than ten seconds with these dry glass bead sterilizers. Glass beads are heated to 250°C to effectively destroy all microbes and spores. Suitable for sterilizing scissors, inoculating loops, and needles.
Sterilizers have a small footprint and can be conveniently placed on the benchtop. Power consumption is low allowing for all day operation. Dial thermometer on front of sterilizer ensures proper heating and back-up safety thermostat prevents over-heating.
What's Included:
one 10 oz bag of glass beads, 6-ft cord and U.S. Standard plug (110 VAC models) or European plug (220 VAC models).
Specifications
Product Type
Sterilizer
Chamber size (" D)
5
Chamber size (" Dia)
1.75
Max temperature (º C)
250
Dimensions (" W)
5
Dimensions (" H)
8
Dimensions (" D)
5.5
Dimensions
Overall: 5"W x 5-1/2"H x 5-1/2"D
CE Compliance
Yes
Temp uniformity
±5°C
Power (VAC)
220
Power (Hz)
50
Power (watts)
175
Materials of construction
Rust resistant chrome-nickel steel
Display
Dial thermometer
Accessories
Qty
/
Price
Availability
Replacement glass beads, 2 mm dia., 10 oz per bag
EW-10779-50
at
$28.50 USD/BAG
In stock.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title