United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
Dual Direction Tensiometer
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-59951-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Tensiometers
>
Tensiometers
Share
|
Dual Direction Tensiometer
EW-59951-25
$5,210.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Ideal for measuring the surface tension of oils, detergents, colloidal suspensions, and chemicals
Measure surface tension in 15 to 30 seconds
Direct-reading scale in dyne/cm
Measure both the surface and interfacial tension of liquids. Range is 0 to 90 dyne/cm. Measurement values are reproducible to within 0.05 dyne/cm. These meters are specified for determining interfacial tension of oil against water by the ring method in ASTM D971, solutions of surface-active agents in ASTM D1331, testing synthetic rubber lattices in ASTM D1417, and determining surface tension of industrial water/wastewater in ASTM D1590. Model 59951-25 measures both upward and downward forces on the measuring ringgreat for testing two-phase liquids. Features graduated dial readings in both directions from 0 to 90 dyne/cm and a double vernier. Both the scales and the corresponding verniers have graduations and figures in different colors to indicate whether the force is in an upward or downward direction.
What's Included:
a 6-cm platinum-iridium measuring ring, three extra torsion wires, and a carrying case.
Specifications
Range
0 to 90 dyne/cm upward/downward direction
Accuracy
±0.05 dyne/cm
Accessories
Qty
/
Price
Availability
Replacement Torsion Ring, for upward/downward direction tensiometer
EW-59951-42
at
$554.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title