Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Dual head piston pump tabletop filler, 220 VAC

EW-76400-15
$7,650.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Dispense solutions for pharmaceutical small batch filling needs with precise accuracy
  • Dispense from <1 to 130 mL
  • Maintain high accuracy (± 0.5% or better) with high cycle rate
  • Handle a wide range of viscosities
Ideal for repetitive dispensing of free-flowing, semi-viscous or viscous fluids.
A rotating micrometer adjustment volume control is attached to the piston pump. The micrometer volume control adjusts to either increase or decrease the desired volume. The longer the piston pump stroke, the greater the dispensed volume. The liquid is drawn in during the first 180 degrees of the rotational motion of the micrometer volume control assembly (intake stroke), and dispensed during the second 180 degrees of the rotation (discharge stroke).
Operate the system via footswitch (included with system) or continuous cycle mode. Use a manual nozzle holder with remote switch (order separately) for handheld operation.
Tabletop Filler includes: footswitch, 6-ft. cord with IEC 320 plug
Pump includes: intake and discharge tubing, stainless steel nozzle
Service kit includes: spare O-rings and pump seals
Note: Actual dispense rates may vary depending on product, fill, volume, and accuracy desired.
*Free-flowing=aqueous solutions, water-thin reagents
Semi-viscous=thin oils, conditioner, shampoo, thin syrup, saturated solutions, agar, hand/body lotion
Heavy viscosity=cream, heavy shampoo, paste, detergent, heavy syrup
Product Type Tabletop Piston Pump Filling Stations
Wetted parts SS, silicone, PTFE
Power (VAC) 220

Accessories
Qty/Price
Availability

at $625.00 USD/EACH

Call 1-888-358-4705 for availability.
Build a complete system




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title