Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

EMD Millipore MAS-100 Eco Air Sampler, 100 L/min

MAS  |   Mfr# 1.09227.0001   |   Item# EW-39152-80

$6,330.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Available for order
or call 1-888-358-4705 to order
Qty

  • Results you can trust in monitoring microbial air quality
  • Programmable sampling delay up to 59 minutes eliminating remote control devices
  • Use standard 100mm Petri dishes
Air samplers allow you to monitor microbial air quality. They are used in industries including pharmaceutical, food technology, chemical, industrial, and water and wastewater. Impaction-style SAS air samplers let you collect and quantify airborne bacteria, viruses, molds, and fungi. Some models are ideal for clean room, pharmaceutical, and electronics applications. Others are designed for food testing and processing. MAS air samplers enable excellent recovery of viable particles as small as 1µm. Ensure your air quality meets standards with the selection of air samplers below.The MAS 100 was developed on the Andersen Air Sampler Principle with collection apertures that correspond to Andersen Level 5 for excellent recovery of viable particles as small as 1µm. All models include a built-in calibration reminder that can be set between 1 and 12 months, and calibration is made easy with the new digital anemometer 39182-82 (sold separately). Internal software is easily accessed and controlled using single touch “yes or no?responses and pre-programmed volumes assure reproducible results.

Air flow is 100 liters/minute. The unit and lid cover are both made of anodized aluminum and the lids are autoclavable. Power is supplied via NiMH rechargeable batteries. The display is alpha-numeric LCD, 16 digits with real time and date.

MAS 100EcoTM Air Sampler

Microbiological air sampler for the food & beverage industry
Lightwight units offer an easy, low cost method for air monitoring
Handle doubles as a stand to allow different angles for testing
This model is specifically designed for the food and beverage industry. There are 6 preset volumes: 10, 20, 50, 100, 200 & 500 liters. Users can also reset any of the presets to any volume between 0-1000 liters. Run time for rechargeable NiMH batteries is 3 hours and maximum time for complete recharge is 9 hours.

MAS 100 Air Sampler

Samples up to 45,000 liters of air or 7.5 hours before battery needs to be recharged
This model offers a built-in anemometer compensates for different sampling heads, changes in temperature/pressure, and variably filled petri dishes. Collection volumes of 50, 100, 250, 500, and 1000 liters are preset for selection and can be deactivated for reactivated as needed. A specific volume between 0 and 2000 liters can also be selected.


This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Air Samplers
Qty/ea 1
Manufacturer number 1.09227.0001
Brand MAS

Accessories
Qty/Price
Availability

at $691.00 USD/EACH

Usually ships in 9 days.

at $233.00 USD/EACH

Usually ships in 9 days.

at $420.00 USD/EACH

Available for order
or call 1-888-358-4705 to order

at $338.00 USD/EACH

Available for order
or call 1-888-358-4705 to order




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    5 Questions | 5 Answers
    Displaying questions 1-5
    • Q:

      Can the device be sterilized with ethylene oxide?
      Asked on 7/10/2013 by Jacquie from Minneapolis, MN

      1 answer

      • STAFF EXPERT

        A:

        The unit itself is not able to be sterilized, but the aluminum lid or dust cover can be removed and autoclaved on their own. They are not able to be ETO sterilized.

        Answered on 7/10/2013 by Mike S from Technical Support
    • Q:

      Could this unit me used in an air handling unit or would the air flow be to great?
      Asked on 8/4/2011 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        It may be too much. The internal pump is calibrated to obtain a specific volume of air. You may want to protect the unit from direct air currents to obtain more accurate results.

        Answered on 8/8/2011 by Jen from Technical Support
    • Q:

      Can 90 mm petridishes be used in this air sampler?
      Asked on 4/13/2010 by Smita from india

      1 answer

      • STAFF EXPERT

        A:

        These are designed to use the standard 100mm diameter contact plates. A 90mm contact plate would be a bit too small to get a good fit.

        Answered on 4/14/2010 by Josh from Technical Support
    • Q:

      What is the recommended calibration period and how much does the calibration typicallly cost? Thank you!
      Asked on 2/5/2010 by Anonymous from Chicago, IL

      1 answer

      • STAFF EXPERT

        A:

        We suggest calibrating the unit once a year. For pricing information please contact our Customer Service team; contact information may be found on the website.

        Answered on 2/9/2010 by Jen from Technical Support
    • Q:

      We are thinking about ordering the MAS-100Eco air sampler. What accessories come with this? We would want a carrying case if available, rechargable battery pack, a battery charger, and a tripod. I need to know what already comes with the unit and which accessories I need to order separately.
      thanks
      Richard
      Asked on 6/22/2009 by Anonymous from Fort Myers, Florida

      1 answer

      • STAFF EXPERT

        A:

        The 39152-80 (MAS-100Eco air sampler) comes with 2 NiMH rechargeable batteries, a battery charger, dust cover and instructions.
        Here are the optional accessories:
        Carrying case: 00225WZ
        Rechargable battery pack: 39182-94
        Tripod adapter: 39182-88

        Please contact our customer service representatives or website for additional assistance with pricing and availability.

        Answered on 6/24/2009 by Jaime from Staff Expert
    Displaying questions 1-5

    Do you have a question about this product? 

    Product Reviews