United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
10 >
EW-08700-10
EZ Series Viscosity Cups
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Technical information
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-08700-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Physical Property Testing
>
Viscometers
>
Viscosity Cups
>
EZ Series Viscosity Cups
Share
|
EZ series viscosity cup #2
EW-08700-10
$159.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
5
10%
$143.10/EACH
15
15%
$135.15/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
An easy and economical way to measure viscosity
±3% production tolerance
Easily measure viscosity using an EZ viscosity cup and a stopwatch. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time.
Each stainless steel cup meets dimensions as specified in ASTM D 4212 standards.
Specifications
Product Type
Viscosity Cups
Cup number
2
Range
19 to 156 cst
Brand
EZ
Manufacturer number
EZ2
Model
EZ2
Accessories
Qty
/
Price
Availability
Aluminum carousel stand for EZ series cups
EW-08700-70
at
$397.00 USD/EACH
In stock.
Add to Cart
Cole-Parmer Big-Digit Stopwatch
EW-94460-28
at
$27.00 USD/EACH
In stock.
Add to Cart
Technical Resources
EZ-series viscosity cups conform to ASTM D 816, D 1084, and D 4212.
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
L'affaire est ketchup? Pas si simple que ça...les mesures de viscosité
In the Thick of Things: Measuring Viscosity
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Viscous Fluid Behavior
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title