United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
12 >
Representative photo only
Eagle Acid Storage Cabinet Self Closing Door 60 Gallon
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-09419-01
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Storage and Handling
>
Safety Cabinets
>
Eagle Acid Cabinets
Share
|
Eagle Acid Storage Cabinet, Self-Closing Door, 60 Gallon
EW-09419-01
$1,581.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Epoxy-coated steel for protection from chemical attack
Acid cabinets are constructed of epoxy-powder coated, double-walled, 18-gauge steel with 1-1/2" (3.8 cm) insulating air barrierideal for both flammable and nonflammable acids and corrosive liquids. Cabinets feature dual vents, grounding attachments, and key lock. Includes adjustable HDPE shelf trays for excellent spill containment. Cabinets are blue with hazard label: CAUTION-CORROSIVES.Models with self-closing door have a fusible link that actuates at 160°F (71°C).
Approval:
FM
Compliance:
OSHA/NFPA Code 30
Specifications
Product Type
Acid Storage Cabinets
Capacity (gal.)
60
Color
Blue
Construction
18-gauge steel
Self-Closing
Yes
Shelves
2
Shelf depth (in.)
28"
Compliance
OSHA/NFPA Code 30
Dimensions (" W)
31.25
Dimensions (" H)
65
Dimensions (" D)
31.25
Shpg wt (lbs)
364
Brand
Eagle
Manufacturer number
CRA-6010
Accessories
Qty
/
Price
Availability
Polyethylene Shelf for Eagle Acid Safety Cabinet, 60-gallon
EW-09418-62
at
$134.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Polyethylene Bottom Tray for Eagle Acid Safety Cabinet, 60-gallon
EW-09418-63
at
$90.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Polyethylene Shelf Tray for Eagle Acid Safety Cabinet, 60-gallon
EW-09418-64
at
$96.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Safety Storage Cabinet Usage
Choosing a Safety Cabinet
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title