Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Eagle Flammable Safety Cabinet, Manual-latching, 4 gallon

EW-09418-21
$502.00 USD/EACH

Usually ships in 7 days.
Qty

  • Adjustable steel shelves hold up to 350 lb
Flammable cabinets are constructed of epoxy-powder coated, double-walled, 18-gauge steel with 1½" insulating air barrier—ideal for safely storing flammable liquid containers up to 5 gallons in size. Cabinets feature dual vents, grounding attachments, and key lock. Includes adjustable HDPE shelf trays for excellent spill containment. Hazard label: FLAMMABLE-KEEP FIRE AWAY.Compliance: OSHA, NFPA Code 30 specifications
Product Type Flammable Storage Cabinets
Capacity (gal.) 4
Color Yellow
Construction 18-gauge steel
Self-Closing No
Shelves 1
Shelf depth (in.) 14-3/4"
Compliance OSHA/NFPA Code 30 specifications
Dimensions (" W) 17.50
Dimensions (" H) 22.50
Dimensions (" D) 18
Shpg wt (lbs) 70
Brand Eagle
Manufacturer number 1904





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title