Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Eagle Space Saver Acid Storage Cabinet, Manual-Latching Door, 12 Gallon

EW-09419-05
$667.00 USD/EACH

Usually ships in 7 days.
Qty

  • Epoxy-coated steel for protection from chemical attack
Acid cabinets are constructed of epoxy-powder coated, double-walled, 18-gauge steel with 1-1/2" (3.8 cm) insulating air barrier—ideal for both flammable and nonflammable acids and corrosive liquids. Cabinets feature dual vents, grounding attachments, and key lock. Includes adjustable HDPE shelf trays for excellent spill containment. Cabinets are blue with hazard label: CAUTION-CORROSIVES.

Compliance: OSHA/NFPA Code 30
Product Type Acid Storage Cabinets
Description Space saver
Capacity (gal.) 12
Color Blue
Construction 18-gauge steel
Self-Closing No
Shelves 1
Shelf depth (in.) 14-3/4"
Compliance OSHA/NFPA Code 30
Dimensions (" W) 23
Dimensions (" H) 35
Dimensions (" D) 18
Shpg wt (lbs) 135
Brand Eagle
Manufacturer number CRA-1925





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title