United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Economical Digital Shaking Water Bath 20 L capacity 110 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12052-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Shaking Water Baths
>
Economical Digital Shaking Water Baths
Share
|
Economical Digital Shaking Water Bath, 20 L capacity, 110 VAC
EW-12052-10
$3,460.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Adjustable temperature, stroke length, and speed give these 10- and 20-liter baths the versatility you want
These digital shaking water baths feature a microprocessor-based keypad and digital display with preset time, continuous run, rpm, and temperature. Nonvolatile memory safely stores your instructions during a power outage. Slow-start and stop feature deters splashing and prevents sample loss. Low-water level signals heater shutoff for added sample safety.
Enamel-coated aluminum housing, and seamless, corrosion resistant stainless steel chamber with removable basket are easy to maintain. Baths come with adjustable springs that stretch across the basket to hold racks and other objects in place while the basket shakes. Secure flasks in the basket with industry-standard flask clamps sold separately. Optional gable-type cover (not included) maintains temperature accuracysold separately.
Specifications
Product Type
Water Bath
Bath type
Shaking Water Bath
Capacity (Liters)
20
Min temperature (º C)
Ambient
Max temperature (º C)
100
High-temperature cutoff
None
Temp setting
digital
Low-level cutoff
Yes
Working area (" D)
6
Drain
No drain valve
Wetted materials
304 stainless steel
Heater wattage
1000
Stability
±0.5°C
Power (VAC)
110
Power (Hz)
60
Display
LCD
Accessories
Qty
/
Price
Availability
Cover for 20-L Economical Digital Shaking Water Bath
EW-12052-52
at
$158.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title