United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Economical Submersible Pump High Flow Utility 20 4 GPM 115 VAC 18 cord
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Flow Charts
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-75500-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Submersible Pumps and Sump Pumps
>
Economical Submersible Pumps, High-Flow Utility
Share
|
Economical Submersible Pump, High-Flow Utility, 20.4 GPM, 115 VAC, 18' cord
EW-75500-00
$166.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Versatileoperate pumps in either a vertical or horizontal position
Compact and economical
Screened inlet helps prevent clogging
18 ft (5.5 m) cord length
High-flow submersible utility pumps for use for water transfer with garden hose adpater or recirculation, water displays, air conditioners, and machine tool coolants. Removable inlet screen allows for water removal 1/8" from floor. Oil-filled for longer motor life. UL listed, CSA certified.
Note:
Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
What's Included:
115 VAC models include grounded plug.
Specifications
Product Type
Submersible Centrifugal Pumps
Max flow rate (GPM)
20
Max flow rate (LPM)
75.6
Max head (ft)
26.3
Max head (m)
8.0
Motor (VAC)
115
Motor (Hz)
60
Motor (Amps)
5.0
Motor (hp)
1/6
Motor (type)
Washdown
Max viscosity (cps)
10.7
Wetted materials
Epoxy-coated cast-aluminum, PP, 316 SS, Viton®, nylon, stainless steel, PVC
Max fluid temp (º F)
120
Max fluid temp (º C)
49
Duty cycle
Continuous
Connections (inlet)
screened
Connections (outlet)
1" NPT(F)
Submersible
Yes
Dimensions
6-1/16" L x 4-5/16" W x 7-3/4" H
(15.5 x 10.9 x 19.7 cm)
Accessories
Qty
/
Price
Availability
Automatic switch for pump 75500-00, 115 VAC
EW-75500-50
at
$76.50 USD/EACH
Usually ships in 5 days.
Add to Cart
Technical Resources
CP_Flow_Curve_75500-00.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title