Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Egg-Shaped Stir Bar, 5/8" L x 1/4" Dia; 1/Pk
Item# EW-04768-10
$12.25 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Super-strong alnico V magnets remain coupled to drive magnets—resisting spin out at high stirring speeds.
  • One-piece coating of PTFE resists corrosion, heat, and cold.
  • Chemically inert stir bars won't contaminate the liquid being stirred and are guaranteed to be leakproof.
  • Low-friction PTFE coating ensures smooth spinning and maximum energy transfer.
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.
Product Type Stir bars
Length 5/8"
Diameter 1/4"
For flask sizes 50 to 100 mL
Qty/ea 1
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456