United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-28680-30
Eighteen-position evaporator/concentrator
Click to enlarge
More Product Details
Specifications
Technical Resources
Instruction manual
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-28680-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Evaporators
>
Evaporative Sample Concentrator
>
Cole-Parmer Eighteen-Position Evaporator Concentrator
Share
|
Eighteen-position evaporator/concentrator
EW-28680-30
$3,458.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Economical and reliable evaporation in a compact instrument.
Individual needle valves provide precise control of nitrogen for each sample for fast convenient evaporation
Easily set instrument for a variety of test tube sizes
Rapidly evaporate common organic solvents—ideal for pharmaceutical, environmental, HPLC, GCMS, and general research applications. Individual needle valve controls combined with heat ensure control for fast concentration of multiple samples. Test tube holder securely accommodates test tubes from 8 to 29 mm dia with volumes from 1 to 50 mL. Features coated aluminum, nickel-plated, and stainless steel (SS) parts to withstand corrosive applications. Includes 16-gauge, 4" SS needles for low consumption of N2 gas. Order tubes separately.
Manifold plate pivots 90deg for easy access to needles and test tubes—adjust the height for optimum evaporation. The integrated triple-block heater accelerates sample evaporation.
What’s included: integrated controlled heater, stand assembly, sample
Specifications
Bath temperature
Ambient to 212°F (100°C)
Temp range
ambient to 212°F (100°C)
Dimensions
16"W x 15"H x 11"D
Condenser type
Rack Evaporators
Gas flow control
0 to 5 L/min
Tube size
8 to 25.4 mm dia; 1 to 50 mL
Brand
Cole-Parmer
Power (VAC)
120
Manufacturer number
109A 28680-30
Technical Resources
Instruction Manual.pdf
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title