United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
EW-08901-35
Electronic desiccator with stainless steel shelves
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-08901-35
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-28
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Desiccators
>
Desiccator Cabinets
>
Electronic Desiccator with Built-In Hygrometer
>
Electronic desiccators with stainless steel shelves
Share
|
Electronic Desiccator with Stainless Steel Shelves; interior dim., 11-1/2"W x 12"H x 10"D; exterior dim., 16"W x 12-1/4"H x 12-1/2"D; 230V
EW-08901-35
$2,100.00 USD/EACH
Usually ships in 13 days.
Qty
:
Add to Cart
Save to shopping list
Automatically regenerates desiccant every five hours-no more replacing messy desiccant!
These new electronic desiccators automatically remove moisture from desiccant enclosed in a control box, and regenerate it every five hours. Maintains 25 to 35% humidity level.
Desiccators provide an airtight, dry environment for storing reagents, IR cells, soil samples, drying biological samples, and cooling crucibles. The stainless steel and glass cabinets will not crack, warp, or discolor.
Units are available in two sizes. The 11-1/2" x 12" x 10" size gives you a choice of two flexboard shelves (model 08901-20 and -25) or two stainless steel shelves (model 08901-30, -35). The larger size (model 08901-40, -45) comes with two stainless steel shelves. Shelves are adjustable in half-inch increments. All models include an easy-to-read, door-mounted hygrometer and a 6-ft cord with two-prong plug.
Specifications
Product Type
Cabinet Desiccator
Interior dimensions (" W)
11.5
Interior dimensions (" H)
12
Interior dimensions (" D)
10
Exterior Dimensions (" W)
16
Exterior Dimensions (" H)
12.25
Exterior Dimensions (" D)
12.5
Power (VAC)
230
Power (Hz)
50
Brand
Boekel
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
EW-17030-28
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title