log in or register
Electroporation system, square wave
EW-02894-18
$7,800.00 USD/EACH
Usually ships in 7 days.
Qty
  • Exponential decay, square wave, and electro cell manipulation systems
  • A great alternative to PEG chemical method
  • Increased efficiencies and cell viability
  • Fast and easy-to-use protocols offer improved reproducibility—independent of operator skill
Choose from a comprehensive line of instruments and accessories for both electroporation and electrofusion of mammalian, bacterial, yeast, fungi, insect, and plant cells and tissue. Designed specifically for cutting-edge applications such as adherent cell electroporation, high-throughput cloning, and large-volume electroporation.
This square-wave electroporation system designed for all mammalian electroporation applications. Features finer voltage discrimination and arc quenching. Monitor all key parameters and control pulse intervals. Applications include mammalian cell transfections/gene therapy, mammalian cell protein/drug electroincorporation, nuclear transfer, plant applications, and bacterial and yeast electroporation.
What's included: stated generator or electro cell manipulator; 30 sterile, disposable cuvettes (1-, 2-, and 4-mm gap sizes); cuvette safety stand; and 20-position numbered cuvette rack.
Product Type Electroporation
Description Electroporation system
Applications Electroporation
Wave form square wave
Low voltage mode (Voltage range) 5 to 500 V (1V)
Low voltage mode (Pulse length range) 10 to 999 µsec
High voltage mode (Voltage range) 505 to 3000 V (5V)
High voltage mode (Pulse length range) 10 to 600 µsec
Manufacturer number 45-0002
Accessories
Qty/Price
Availability
at $167.00 USD/PACKAGE
Usually ships in 6 days.
at $167.00 USD/PACKAGE
Usually ships in 2 days.
at $167.00 USD/PACKAGE
Usually ships in 6 days.
at $1,850.00 USD/EACH
Usually ships in 7 days.
at $706.00 USD/EACH
Call 1-888-358-4705 for availability.
at $706.00 USD/EACH
Usually ships in 7 days.
at $772.50 USD/PACKAGE
Usually ships in 7 days.
at $772.50 USD/PACKAGE
Usually ships in 7 days.
at $225.00 USD/PACKAGE
Usually ships in 7 days.
at $225.00 USD/PACKAGE
Usually ships in 7 days.
at $1,381.00 USD/PACKAGE
Usually ships in 7 days.
at $134.00 USD/EACH
Usually ships in 7 days.
at $1,282.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title