United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
16 >
EW-36002-12
Image is representative of item.
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36002-12
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Heating Mantles
>
Electrothermal Electromantles
Share
|
Electrothermal Electromantle, 500-mL capacity, 115 VAC
EW-36002-12
$556.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Features a durable, chemical resistant polypropylene housing that is "safe to the touch".
Flexible heating element ensures minimum risk of flask breakage.
These controlled and grounded electromantles feature a built-in power regulator and safety earth (ground) screen. Unique ventilation slots keep the exterior "safe to the touch" even while in use.
The replaceable coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support. Ceramic fiber insulation keeps the chemical-resistant polypropylene case cool. Built-in power controller has dial markings from 1 to 10 for repeatability. Mantles feature indicator lights: an amber-colored "power on" light and a clear-colored "heater on" light. All mantles are double fused and include a single support rod clamp that accepts a 1/2" rod. The stainless steel grounding screen provides added protection against shock in the event of spillage or flask breakage. Mantles include 6 1/2-ft (2-m) power cord; the 115 VAC models include a U.S. plug (230 VAC models do not include plugs). Call our Application Specialists to order replacement heating element cartridges.
Specifications
Product Type
Heating Mantle
Watts
200
Flask size (mL)
500
Max temperature (º F)
842
Max temperature (º C)
450
Dimensions
12.167 in W x 5.667 in H x 9.33 in D
Power (VAC)
115
Brand
Electrothermal
Manufacturer number
EM0500/CEX1
Model
EM0500/CEX1
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Keeping Samples the Right Temperature
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title