United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
Electrothermal MM Series Micro Kjeldahl Extraction Heater 600 W 230V 50 60Hz UK plug
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-03074-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Extraction Apparatus
>
Electrothermal MM Series Micro-Kjeldahl Extraction Heater
Share
|
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 230V/50/60Hz, UK plug
EW-03074-32
$2,330.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Specifically designed to determine the protein content of food on a micro scale - from 18 to 50 mL
6 positions allow single or multiple digestions
Rugged stainless steel construction minimizes damage from spills and is easy to clean
Dedicated controller for each mantle is safely housed in a "cool zone"
This six-position extraction heater is designed for Kjeldahl digestion on a micro scale - accommodating up to six (6) round-bottom flasks between 18 to 50 mL in size. Each mantel has an independent heater with control dial, ON/OFF switch, and indicator light, and operates up to a maximum element temperature of 800°C.
The lower part of the unit containing the control equipment is safely separated from the upper heating elements by a well-ventilated air space. Unit is double fused on the supply line and neutral for extra safety.
What's Included:
support arms with brackets and a 6-ft (1.8-m) power cord
Specifications
Product Type
Extraction Heater
Watts
600
Recesses
6
Flask max capacity (mL)
50
Flask min capacity (mL)
18
Max element temperature range (°C)
800
Min element temperature range (°C)
550
Case
Stainless Steel
Thermal insulation
Ceramic fiber/mineral wool
Dimensions (D x W x H) (cm)
16 x 52 x 16.2
Dimensions (D x W x H) (")
6-3/8 x 20-1/2 x 6-5/16
CE Compliance
Yes
Plug type
UK
Power (VAC)
230
Power (Hz)
50/60
Brand
Electrothermal
Manufacturer number
MM2313/E
Model
MM2313/E
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title