dummy
EW-19505-05
- Improved user interface allows direct access to calibration mode and set point adjustment
- Universal power supply input from 85 to 250 VAC/DC allows use at multiple sites
- Automatic temperature compensation ensures accurate measurements
- Galvanically isolated 4 to 20 mA output for logging data
These 1⁄8-DIN controllers display the measured parameter value or temperature with a touch of a button. The user interface was simplified for quick set point review and adjustment and entrance into the calibration mode by simply pressing keys on the front panelno need to remember and enter security codes.
The controller has two relays that can be set for any combination of low and/or high level control. Adjustable temperature coefficients and hysteresis provide accurate control in varying applications. Select from any of the six measurement ranges for desired resolution and accuracy.
| Dimensions |
3-3/4"W x 1-7/8"H x 6"D |
| Control type |
On/Off |
| Display |
4-digit, 14-segment LED |
at $169.00 USD/EACH
Call 1-888-358-4705 for availability.
at $176.00 USD/EACH
Call 1-888-358-4705 for availability.
at $112.00 USD/EACH
Call 1-888-358-4705 for availability.
at $173.00 USD/EACH
In stock.
at $173.00 USD/EACH
In stock.
at $171.00 USD/EACH
Usually ships in 9 days.
at $151.00 USD/EACH
In stock.
at $228.00 USD/EACH
Call 1-888-358-4705 for availability.
at $39.00 USD/EACH
In stock.
at $115.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection