United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-56700-00
Eutech Instruments pH 200 1/8-DIN pH/ORP controller
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Reconditioned & Like New
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-56700-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-20
NIST-Traceable Calibration; pH Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/8-DIN pH/ORP Control
>
Eutech Instruments pH 200 1/8-DIN pH/ORP Controller/Transmitter
Share
|
Eutech Instruments pH 200 1/8-DIN pH/ORP controller
EW-56700-00
$449.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Economical controller is designed with the features and reliability of a much more expensive instrument.
Two-level password protection allows calibration access without accidentally changing controller set-up.
Symmetrical or asymmetrical mode operation
Tactile, water-resistant membrane keypad
This compact and economical controller has removable terminal strips and accepts power input between 85 to 250 VAC/DC for simple installation. The scrolling 14-segment LED display guides the user through setup, calibration, and operation functions.
Setup and calibration modes are password-protected to prevent unauthorized changes. Operator can review previous calibration points and the electrode slope and offset for diagnostic purposes. The controller can be set for automatic or manual temperature compensation depending upon the application. The user can select one- or two-point calibration at seven common buffer values.
Two independently programmable relays can be customized for set points and hysteresis. LED enunciators on the front panel indicate operation mode, measured parameter, and relay status. A galvanically isolated, 4 to 20 mA output is available for logging purposes. Order 100Ω Pt RTD, tinned-end electrodes with BNC pH connectors separately.
What's Included:
Mounting assembly and instructions.
Specifications
Product Type
Controller
Range (pH)
0.00 to 14.00
Range (Temperature)
-10 to 110°C
Range (mV)
-1000 to 1000
Accuracy (pH)
-0.01 to 0.01
Accuracy (Temperature)
-0.5 to 0.5°C
Accuracy (mV)
-2 to 2 mV
Resolution (pH)
0.01
Resolution (Temperature)
0.1°C
Resolution (mV)
1 mV
Output (Current)
4 to 20 mA, 200 ohm max load
Output (Relay)
Two SPDT (120 VAC, 6 A max; 240 VAC, 3 A max
Dimensions
3.75 in W x 1.875 in H x 6 in D (9.6 cm W x 4.8 cm H x 15.2 cm D)
Control type
On/Off
Password protection
Yes
Display
4-digit LED
Power
110/220 VAC, 50/60 Hz (adapter included)
Housing
1/8-DIN
Brand
Eutech Instruments
Accessories
Qty
/
Price
Availability
Benchtop stand for 1/8 DIN panel meters
EW-05656-55
at
$17.00 USD/EACH
In stock.
Add to Cart
120 VAC Power Cord With U.S. Standard Plug, 6-ft (1.8-m) Long
EW-50001-00
at
$39.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
EW-17106-20
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
LIKE NEW 56700-00
EW
-
00308RX
at
$363.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (366 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
pH/ORP Control Systems Selection Guide
Typical Problems in Industrial pH Measurement & Control
NEMA/IEC Enclosure Ratings
Show All...
Taking Control of Your pH!
Tech Challenge: Faulty pH Readings
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title