United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-56717-32
Eutech Instruments
®
pH 560 pH/orp Monitor With Two Relays
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-56717-32
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-20
NIST-Traceable Calibration; pH Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
Eutech Instruments 1/2-DIN pH/ORP Control
>
Eutech Instruments pH 560 1/4-DIN pH/ORP Controller
Share
|
Eutech Instruments pH 560 pH/orp Monitor With Two Relays
EW-56717-32
$356.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Small size, economical monitor or controller with display
Measure and display both pH/ORP and temperature
Automatic or manual temperature compensation
Asymetrical or symmetrical mode operation
The pH 550 and 560 monitors are used to measure pH /ORP and temperature values in applications such as water treatment and monitoring, chemical processing, or food processing. The monitors have user-friendly features like menu-driven programs and calibrations that are easily performed with the push-button keypad. All setup and calibration parameters are retained in the nonvolatile memory to prevent data loss during power outages.
Measurements are compensated for temperature variations automatically when used with pH electrodes that have temperature elements. The monitors also offer manual temperature compensation with independent settings for calibration and process temperature. One- or two-point calibrations can be performed with USA or NIST buffer sets. All values, operational, and error messages are indicated on the large, dual-line LCD. Order 100
Pt RTD pH and ORP electrodes from pages XXXX-XXXX. Order BNC to spade lug adapter to install pH or ORP electrodes.
The pH 560 monitor has additional features including two switching contacts as set point relays. The relays have separately adjustable high and low set point hysteresis (dead bands) to prevent relay chattering. The relays feature a 0 to 2000 second time delay to prevent false alarms and unneeded control. The pH 560 also features a backlit and UV light protected display.
What's Included:
110 VAC to 9 VDC power adapter
Specifications
Product Type
Controller
Range (pH)
0.00 to 14.00
Range (Temperature)
-10 to 125°C
Range (mV)
-1000 to 1000
Accuracy (pH)
-0.01 to 0.01
Accuracy (Temperature)
-0.5 to 0.5°C
Accuracy (mV)
-2 to 2 mV
Resolution (pH)
0.01
Resolution (Temperature)
0.1°C
Resolution (mV)
1 mV
Output (Relay)
Two SPDT
Dimensions
3.75 in W x 3.75 in H x 2.833 in D (96 x 96 x 66 mm)
Control type
On/Off
Relay delay
Programmable 0 to 2000 seconds
Dead band
Programmable
Display
LCD
Power
110/220 VAC to 9 VDC (adapter included)
Housing
IP65
CE Compliance
Yes
Brand
Eutech Instruments
Accessories
Qty
/
Price
Availability
BNC-to-stripped wire ends connector
EW-56105-60
at
$14.28 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
EW-17106-20
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual (1040 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
pH/ORP Control Systems Selection Guide
Typical Problems in Industrial pH Measurement & Control
NEMA/IEC Enclosure Ratings
Show All...
Taking Control of Your pH!
Tech Challenge: Faulty pH Readings
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title