log in or register
Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
EW-28690-50
$999.00 USD/EACH
Usually ships in 12 days.
Qty
  • Indivdual needles provide equal flow of gas for faster, consistent evaporation of samples
  • Economical and reliable evaporation in a compact instrument
  • Models available to accommodate different-sized tubes and microwell plates
Rapidly evaporate common organic solvents—ideal for pharmaceutical, environmental, HPLC, GCMS, and general research applications. Needle valve controls the flow of gas into the equalizer manifold, which provides equal gas flow and uniform evaporation to each well. Needle valves ensure control for fast concentration of multiple samples. Each needle is made from 15-gauge stainless steel with easy height adjustment to accommodate a variety of sample sizes.Stand-Alone Evaporator/Concentrators are made of autoclavable polypropylene with a nickel-plated valve; feature cutouts for easy removal of plates. Select either a 96- or 384-well model.
Total positions 96
Bath temperature Ambient
Temp range ambient
Condenser type Stand-Alone Evaporators
Gas flow 1 to 100 mL/needle
Brand Cole-Parmer
Manufacturer number 099AEV96
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title