United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-50510-75
Extech HD600 : Data Logging Sound Level Meter
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-50510-75
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-53
NIST-Traceable Calibration; Sound Meter (Type II)
$185.00 USD/EACH/EACH
Usually ships in 6 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Sound Meters
>
Extech Data Logging Sound Level Meter
Share
|
Extech HD600 : Data Logging Sound Level Meter
EW-50510-75
$559.99 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
High accuracy datalogging capability of up to 20,000 records at speeds from 1/sec to 1/59sec
Meets ANSI and IEC 61672-1 Type 2 standards
Three ranges from 30 to 130dB with 1.4dB accuracy
Data logging capability up to 20,000 records
Displays and records readings with real date and time stamp
Captures up to 10 reading/sec when connected to a PC
Min/Max, Data Hold, Auto power off functions
Heavy duty rugged double molded housing with built-in stand
Built-in USB port includes software
What's Included:
Complete with AC adaptor, Windows® compatible software, USB cable, wind cover, tripod, 9V battery, and hard carrying case
Specifications
Product Type
Sound Meter
Meter class
Type 2
Decibel (dB) range (high)
130
Decibel (dB) range (low)
30
Compliance
ANSI/IEC type 1 and type 2
Function
Min/Max, hold
Accuracy
±1.4dB
Weighting
A and C
Resolution (dB)
0.1 dB
Response
Fast/slow time
Display
LCD
Datalogging
20,000 points
Output
AC/DC (analog)
Brand
Extech
Dimensions
10.9 x 3 x 2 in (278 x 76 x 50mm)
CE Compliance
Yes
Power
One 9V battery
Manufacturer number
HD600
Model
HD600
Accessories
Qty
/
Price
Availability
Tripod
EW-90415-32
at
$29.99 USD/EACH
Usually ships in 6 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Sound Meter (Type II)
EW-17101-53
at $185.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
More info
Tech Tips
Specification Sheet
Related technical articles (Roll over for summary)
Sound Meter Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title