United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-37803-14
Extech Humidity/temperature Pen
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-37803-14
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-28
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Humidity Products
>
Humidity Meters / Thermohygrometers Meters
>
Pen-style Thermohygrometer
>
Extech Thermohygrometer Pen
Share
|
Extech Humidity/temperature Pen
EW-37803-14
$33.99 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Pocket size with quick and reliable measurements
Pen displays relative humidity and temperature simultaneously. Min/max function displays and holds the maximum humidity and temperature values since power-up in memory; simply press the clear button to reset. Featuring an on/off button, and selectable °F/°C temperature.
What's Included:
one pocket clip and battery.
Specifications
Product Type
Thermohygrometer
RH range
20 to 90%
RH accuracy
±5%
RH resolution
1%
Max temperature (º C)
50
Temp accuracy
±2°F (±1°C)
Temp resolution
0.1°F (0.1°C)
Response time
8 seconds
Display
LCD
Max temperature (º F)
122
Min temperature (º F)
14
Min temperature (º C)
-10
Temp range
14 to 122°F
(-10 to 50°C)
RH range (Max)
90%
RH range (min)
20%
CE Compliance
Yes
Dimensions
1 in W x 6 in H x 0.75 in D
Power
on LR44 battery (included)
Brand
Extech
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
EW-17030-28
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
More info
Tech Tips
Specification Sheet
Related technical articles (Roll over for summary)
Humidity Expressed as Dewpoint Temperature
Temperature FAQs
Temperature Instrument Ranges
Show All...
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title