log in or register
Extech MO290 InspectorPro 8-in-1 Combination Pin and Pinless Moisture Meter
EW-59820-70
$529.00 USD/EACH
Usually ships in 7 days or sooner.
Qty
  • Measure moisture, humidity, and temperature via a built-in probe or non-contact infrared.
  • Pinless moisture sensor allows to monitor with no surface damage
  • Select from multiple wood types and measurement ranges
  • Non-Contact 8-to-1 Infrared Thermometer
  • Built-in thermohygrometer
The InspectorPro moisture meters quickly indicate the moisture content to a depth of 0.75" (19mm) below the surface of materials using pinless technology; or you can use a remote pin-type probe on materials that require penetration to get a reliable reading. Each meter can simultaneously display moisture and air temperature, IR temperature, or humidity. The built-in thermohygrometer measures relative humidity, air temperature plus grains per pound, dew point, vapor pressure, and condensation point. The infrared thermometer measure non-contact surface temperature with a 8:1 distance to spot ratio and has a 0.95 fixed emissivity. Advanced features include an easy to read, large dual display with automatic backlight, programmable high/low alarms, differential temperature, Min/Max, data hold, auto power off and low battery indicator.
InspectorPro MO295 also includes a built-in memory that can store up to 20 labeled readings for further analysis and includes analog bar-graph.
Water Restoration Package MO290-RK includes essential probes to measure moisture levels under baseboards, in between walls, and in hard wood and plywood subfloors. The kit also comes with a RHT10 humidity/temperature datalogger that can store up to 16,000 readings and Includes software for downloads to PC. Software can convert readings to Grains per Pound.
Optional pined probes can be ordered below to meet a specific application.
What's Included:
pin moisture probe with cable, 9V battery and carrying case
Product Type Pin and Pinless Moisture Meters
Resolution 0.1, 0.1F/C
Measurement Type Combination
Moisture range 0 to 99.9 (Relative)
Alarm Yes
Display Digital
Dimensions (W x H x D) 6.5 x 2.8 x 1.5inc (165 x 70 x 38)
Power One 9 V battery (included)
CE Compliance Yes
Brand Extech
Manufacturer number MO290
Model MO290
Make InspectorPro
Accessories
Qty/Price
Availability
at $139.99 USD/EACH
Usually ships in 7 days.
at $84.99 USD/EACH
Usually ships in 7 days.
at $84.99 USD/EACH
Usually ships in 7 days.
at $69.99 USD/EACH
Usually ships in 7 days.
at $84.99 USD/EACH
Usually ships in 7 days.
at $84.99 USD/EACH
Usually ships in 7 days.
at $59.99 USD/EACH
Usually ships in 7 days.
at $69.99 USD/EACH
Usually ships in 6 days.
at $69.99 USD/EACH
Usually ships in 7 days.
Replacement parts
Qty/Price
Availability
at $25.60 USD/PACKAGE of 4
In stock.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title