United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
EW-33704-02
EXTRACT-ALL
PORTABLE AIR CLEANING SYSTEM WITH 4' TO 6' LONG EXTRACTOR ARM, 120 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33704-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Furniture & Fume Hoods
>
Fume Extractors
>
Extract-All Portable Air Cleaning System
Share
|
Extract-All Portable Air Cleaning System with 4' to 6' Long Extractor Arm, 120 VAC
EW-33704-02
$3,942.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Multi stage filtration
Durable cabinet construction
Convenient handle and casters provide easy mobility
Highly efficient, yet low-cost replacement filters
Quiet operation
Long life blower motor
Adjustable extractor arm with fire-retardant properties
Portable air cleaning system is designed for direct source-capture removal of smoke, dust, fumes, and other airborne contaminants from a wide variety of industrial and other applications. Use to improve the air quality in your facility by effectively removing airborne contaminants beforre they enter the user''s breathing zone. Easily place adjustable extractor with aluminum hood where you need work done.
What's Included:
Four filters, three that are replaceable
Specifications
Brand
AIR IMPURITIES REMOVAL SYSTEM
Accessories
Qty
/
Price
Availability
Secondary Merv 8 Pleated Filter
EW-33704-90
at
$56.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Primary Merv 14 Cell Filter
EW-33704-91
at
$186.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Activated Carbon After-Filter
EW-33704-92
at
$90.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title