United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 16
of
18 >
Representative photo only
Federal Signal Low Profile Strobe Light Surface mount Amber
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10402-35
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Alarms
>
Federal Signal Low Profile Strobe Light
Share
|
Federal Signal Low Profile Strobe Light, Surface mount , Amber
EW-10402-35
$69.86 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Long life in a compact strobe light
NEMA 4X/IP65-rated, low-profile strobes are ideal for mounting on control panels and other flat or flush surfaces. Strobes operate in temperatures from -31 to 150°F (-35 to 66°C). Threaded screw-on lens provides tool-free wiring and strobe tube replacementstrobe tube rated for 7000 hours of use. High-quality, long life strobe lamps are designed to reduce tungsten build-up for longer lamp maintenance cycles. Strobe mechanism produces 2.2 joules of energy while drawing low amperage. Strobes have a flash rate of 75 to 85 flashes per minute. Availablein three mounting configurations: 1/2" NPT(M) pipe mount with 18" wire leads, 1/2" NPT(F) pipe mount, and 3-hole surface mount.
Specifications
Product Type
Strobes - surface-mount
Power (VAC)
120
Operating conditions
-31 to 150°F (-35 to 66°C)
Enclosure
NEMA 4X / IP65
Dimensions
5"H x 3-1/8" dia
Brand
Federal Signal
Manufacturer number
LP3S-120A-CP
Model
LP3S-120A-CP
Technical Resources
Related technical articles (Roll over for summary)
Alarm/Status Indicators: An Overview
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title