United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-54100-02
Fischer Technology FMP 20 Dualscope
®
Coating Thickness Gauge
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-54100-02
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-69
NIST-Traceable Calibration; Coating Thickness Gauge (mechanical type)
$99.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Thickness Gauges
>
Fischer Precision Coating Thickness Instruments
Share
|
Fischer Technology FMP 20 Dualscope Coating Thickness Gauge
EW-54100-02
$955.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Available for sale in the United States, Canada, and Mexico only
Portable instruments deliver non-destructive, high precision measurements
All models feature:
Automatic probe recognition
USB port for data transfer to a PC
Large display rich in contrast - 240x160 pixels
Memory for up to 1000 readings
Audible signal at measurement acquisition
Displays mean value, standard deviation, min, max, and range
Easy adaptation to the shape of the specimen
Additional corrective calibration using one or two calibration foils
Master calibration for exact characteristics settings for extreme material and geometric properties
Capable of storing the master calibration in the connected probe
Units of measurement can be switched between µm and mils
Adjustable instrument switch-off or continuous operation
Low battery indication
Lockable keyboard/restricted operating mode
Mechanical sliders to cover keys not required for the measurement operation
Various language settings
These easy to operate, rugged instruments are suitable for virtually any coating thickness measurement requirement. With a wide assortment of plug-in type probes these meters are designed for non-destructive coating thickness measurements according to the magnetic induction method (Deltascope
®
FPM10), the eddy current method (Isoscope
®
FPM10), or both methods in one instrument (Dualscope
®
FPM20). In addition to one user created calibration application, the spectrum of requirements that this series is capable of spans from extensive painted specimens to other very specific and complex measuring challenges.
Deltascope FMP10
Use for the measurement of non-ferromagnetic metal
Coatings such as chrome, copper, zinc, as well as paint,
lacquer, enamel, or plastic coatings on steel and iron.
Isoscope FMP10
Use for the measurement of paint, lacquer or plastic
coatings on non-ferromagnetic metal substrate materials.
Use for anodic coatings on aluminum as well as electrically
conducting coatings on non conducting carrier materials.
Dualscope FMP20
Includes automatic substrate material recognition feature
for the combination of both measurement methods. This
universal instrument is capable of measuring numerous
coatings both on iron/steel, non-ferromagnetic metals,
and on non-conducting carrier materials.
What's Included:
USB interface cable, carrying strap, and four AA batteries.
Specifications
Product Type
Precision Coating Thickness Measurement Systems
Display
240x160 pixels
Description
Dualscope FMP20 coating thickness gauge
Dimensions
7 in L x 3.5 in W x 1.5 in H (17.7 x 8.8 x 3.8 cm)
Power
Four AA batteries (included); optional AC adapter (advanced models)
Brand
Fischer Technology
Model
604-285
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Right angle, single tip, Model FGABW1.3, 0 to 60 mils, for models Deltascope FMP10 & Dualscope FMP20
EW-54100-06
at
$745.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Probe for High impact, shatter resistant, Model FTA3.3H, 0 to 45 mils, for models Isoscope FMP10 & Dualscope FMP20
EW-54100-08
at
$645.00 USD/EACH
In stock.
Add to Cart
Right angle, single tip, 0 to 45 mils, Model FAW3.3, for Isoscope FMP10 & Dualscope FMP20
EW-54100-10
at
$865.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Magnetic Induction Probe, Pencil type, 0 to 98.43 mils
EW-54102-23
at
$495.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Coating Thickness Gauge (mechanical type)
EW-17101-69
at $99.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title