Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Home  >    
Fischer Technology MP20E-S, Coating Thickness Dualscope Substrates
Fischer Technology  |   Item# EW-59971-28
$795.00 USD/EACH
Discontinued
Qty
  • Available for sale in U.S., Canada, and Mexico only
  • Easily exchangeable plug-in probes
  • All magnetic induction, eddy current and dual probes can be connected
Application-specific probes are the key to successful coating thickness measurement. Probes are the most important component of systems used to measure coating thickness.
Select magnetic induction or eddy current method probes that best meet your application. Fischer probes are adapted and optimized for applications; they are long lasting and reliable.
Probes work with the Fischer Digital Coating Thickness Measurement Dualscope MP20 - which offers an economic solution for the measuring of coatings / platings on ferrous and / or non-ferrous substrate materials.
Probes also work with the Dualscope MP40 - which combines the magnetic induction and eddy current methods in one hand-held instrument, similar to the MP20. The combination enables the user to measure in random order paint, plastic, and organic coatings on non-ferrous metals and on steel, and non-ferrous metal coatings on steel with one and the same instrument.
Display LCD
Model 603-693/E
Brand Fischer Technology
Accessories
Qty/Price
Availability
at $662.00 USD/EACH
Usually ships in 7 days or sooner.
Calibrations
Qty/Price
Availability
at $124.00 USD/EACH
Usually ships in 6 days.
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456