United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
Fixed Snubber 304 316 SS 5000 psi Water or Light Oil
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68800-82
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Gauge guards, Seals, and Accessories
>
Pressure Snubbers
>
Fixed 316 SS snubbers
Share
|
Fixed Snubber, 304/316 SS, 5000 psi, Water or Light Oil
EW-68800-82
$16.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Snubbers eliminate needle fluctuations from pressure pulsations—extends gauge life. Gauge connection is 1/4'' NPT(F); process connection is 1/4'' NPT(M). Use optional bushings to adapt snubbers or gauges.
Fixed snubbers feature an element made of micro-sized stainless steel pieces, sintered to a porosity of 10 microns for use with a specified fluid (see table below). Adjustable pressure snubbers can be used with a variety of fluids and conditions; feature a tunable choke valve.
Specifications
Product Type
Pressure gauge accessories
Range
0 to 5000 psi
Process connection
1/4" NPT(M)
Accessories
Qty
/
Price
Availability
Threaded reducing bushing, PFA, 1/2" NPTM x 1/4" NPTF
EW-08539-85
at
$17.00 USD/EACH
Usually ships in 14 days.
Add to Cart
Reducing bushing, 1/4
EW-08539-89
at
$16.50 USD/EACH
In stock.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title