log in or register
Fixed Snubber, Brass, 1500 psi, Air
EW-68800-70
$12.00 USD/EACH
Usually ships in 9 days.
Qty
    Snubbers eliminate needle fluctuations from pressure pulsations—extends gauge life. Gauge connection is 1/4'' NPT(F); process connection is 1/4'' NPT(M). Use optional bushings to adapt snubbers or gauges.
    Fixed snubbers feature an element made of micro-sized stainless steel pieces, sintered to a porosity of 10 microns for use with a specified fluid (see table below). Adjustable pressure snubbers can be used with a variety of fluids and conditions; feature a tunable choke valve.
    Product Type Pressure gauge accessories
    Range 0 to 1500 psi
    Process connection 1/4" NPT(M)
    Accessories
    Qty/Price
    Availability
    at $17.00 USD/EACH
    Usually ships in 14 days.
    Reducing bushing, 1/4
    EW-08539-89
    at $16.50 USD/EACH
    In stock.
    Technical Resources
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title