United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 14
of
20 >
EW-44765-65
Flammable Material Undercounter Refrigerator/Freezer, 6 cu ft
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-44765-65
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Refrigerators
>
Refrigerator and Freezer Combination
>
Marvel Refrigerators & Freezers
Share
|
Flammable Material Undercounter Refrigerator/Freezer, 6 cu ft
EW-44765-65
$3,103.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Durable commercial cooling at an affordable price
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.
Flammable Materials Models
are designed to completely insulate the storage compartment from chance of electrical sparking. The mechanical assembly is designed to prevent electrical sparks from igniting volatile or explosive substances in the surrounding environment. These products are listed by UL File SA7488(N).
What's Included:
Commercial power cord and plug, except for explosion-proof models which require hard-wiring.
Specifications
Product Type
Undercounter Combos
Capacity (cu ft)
6.1
Capacity (Liters)
173
Dimensions (interior)
28"H x 21"W x 20 3/4"D
Dimensions (overall)
34"H x 23 7/8"W x 26"D
Min Temp - Refrigerator (º C)
1
Min Temp - Refrigerator (º F)
33
Max Temp - Refrigerator (º C)
7
Max Temp - Refrigerator (º F)
45
Min Temp - Freezer (º C)
-18
Min Temp - Freezer (º F)
0
Max Temp - Freezer (º C)
-9
Max Temp - Freezer (º F)
15
No. of doors
1
No. of shelves
2 interior, 3 door
Power (Hz)
60
Brand
Marvel
Manufacturer number
6FRF0001
Power (VAC)
115
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title