log in or register
Flare compression pipe adapter, 1/2" x 1/8", PTFE, Each
EW-30490-34
$48.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Leak tight & high-pressure connection for corrosive chemical applications
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leak-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing. PFA has a low coefficient of friction and therefore does not form a tight seal with standard gripper fittings. Flare fittings are a great alternative to gripper fittings in critical applications. To install Flare fittings you slide the flare nut on to a piece of PFA tubing, the tubing is then heated using a heat gun and then a mandrel is inserted in the fitting to create the flare. The flared end of the tubing is then inserted into the body of the fitting and the nut tightened down on the flare. Choose from the various styles. If there is another type or style not show call our Application Specialists for more information on addition fitting types.
Product Type Compression fittings
Material PTFE
Quantity 1
Brand Parker Hannifin
Manufacturer number FAMS-82
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title