United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
14 >
Representative photo only
Flare compression pipe adapter 3 8 x 3 8 PTFE Each
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-30490-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fittings
>
Flare Fittings
>
Flare to NPT threaded straight adapters
Share
|
Flare compression pipe adapter, 3/8" x 3/8", PTFE, Each
EW-30490-30
$44.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Leak tight & high-pressure connection for corrosive chemical applications
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leak-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing. PFA has a low coefficient of friction and therefore does not form a tight seal with standard gripper fittings. Flare fittings are a great alternative to gripper fittings in critical applications. To install Flare fittings you slide the flare nut on to a piece of PFA tubing, the tubing is then heated using a heat gun and then a mandrel is inserted in the fitting to create the flare. The flared end of the tubing is then inserted into the body of the fitting and the nut tightened down on the flare. Choose from the various styles. If there is another type or style not show call our Application Specialists for more information on addition fitting types.
Specifications
Product Type
Compression fittings
Material
PTFE
Quantity
1
Brand
Parker Hannifin
Manufacturer number
FAMS-66
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title