log in or register
Flaring tool for tubing 1/4 to 1" outer diameter
EW-30490-01
$360.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
    Use these tools for a better fit. Heat the PFA tubing with a heat gun, and insert the tool to create the flare. Tools are available for any size of tubing from 1/4-inch to 1-inch outer diameter.
    Product Type Compression fittings
    Quantity 1
    Brand Parker Hannifin
    Manufacturer number MFT-U
    Accessories
    Qty/Price
    Availability
    at $142.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $147.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title