United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
5 >
Representative photo only
Flexible small diameter fiberglass insulated heating cord 3 53 watts 120 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-03122-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Heating Tapes, Cords, Blankets
>
Heating Cords - Fiberglass / Flexible Small-Diameter
>
Flexible Small-Diameter Fiberglass Heating Cords - 120 VAC
Share
|
Flexible, small-diameter, fiberglass insulated heating cord, 3', 53 watts, 120 VAC
EW-03122-30
$100.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Heat to 850ºF (454ºC)
Only 3/16" diameter-ideal for heating in tight spaces
Spiral wound heating element-packs more heating element into each inch of cord.
Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications.
A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord. Cords feature low power consumption: the 3-,6-, and 12-ft 120 VAC cords (heating 1/4", 1/2", and 1" glass tubing respectively) draw about 0.1 watt/inch at full voltage/high temperature.
Control heating cord temperature with our DIGI-SENSE
®
temperature controller model 89000-00 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads. The 120 VAC models have tip plugs and require the power cord adapter 03122-71; 240 VAC models have stripped ends.
Use this table to determine the best coil spacing for you application. For example, to achieve a maximum temperature of 602 ºF (317ºC) on a 1" OD tubing at 120 VAC, you need to space the heating coils 1/4" apart.
Note: Readings were taken on an open-end glass tube with no forced air circulation; glass tubing had 1/16" wall. A power controller is needed to adjust voltages below 115 VAC.
Warning: Do not overlap heating cords!
Specifications
Product Type
Cords
Watts
53
Power (VAC)
120
Length (ft)
3
Max temperature
900°F (482°C)
Accessories
Qty
/
Price
Availability
Steel cased variable-voltage controller 3PN1010B; 120 VAC in, 0 to 140 V out, 10 A
EW-01575-00
at
$382.00 USD/EACH
In stock.
Add to Cart
Power Cord Adapter
EW-03122-75
at
$39.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
How to Easily Install Heating Tape
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title