United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Flourescent FLT Yellow Green Dye Concentrate Pint
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-00298-17
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Dye Tracers
>
Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Share
|
Flourescent FLT Yellow/Green Dye Concentrate, Pint
EW-00298-17
$26.50 USD/PINT
In stock.
Qty
:
Add to Cart
Save to shopping list
Determine water flow in closed circuits or open channels
Biodegradable and nontoxic
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
Specifications
Product Type
Dye Tracers
Gallons of water for 1 ppm
12,500
Applications
Squirt-top dispensing
Manufacturer number
106501P
Technical Resources
00298-17MSDS.pdf
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title