log in or register
Flourescent FWT Red Dye Powder, 1 Lb
EW-00298-26
$144.00 USD/EACH
  • Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
  • Image may not represent dye color
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent red dye (rhodamine WT) is noncarcinogenic. Ideal in yellowed water or water with heavy algae growth. Use an ultraviolet lamp (order separately) to trace fluorescent red dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
Product Type Dye Tracers
Gallons of water for 1 ppm 60,400
Applications Cooling towers, lakes, rivers, ponds, power plants
Manufacturer number 105503
Technical Resources
  • MSDS RED DYE
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title