United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-10322-30
FlowMarker Fog Generator
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10322-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Clean Room
>
Airflow Measurement
>
FlowMarker Fog Generator
Share
|
FlowMarker Fog Generator
EW-10322-30
$6,329.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Visualize natural air currents without creating disturbances
Portable design allows operation in hard-to-reach areas
Nontoxic fog breaks down into carbon dioxide and water
Identify potential dead spots in room airflow patternsthe FlowMarker marks air movements through critical environment areas, fume hoods, biosafety cabinets, HVAC systems and low air velocity applications. The fog consists of FDA-approved, non-toxic polyfunctional alcohol preparation, which meets standards for USP purity (Food grade), and highly purified demineralized water. When full, the 16-mL fluid reservoir allows operation for 20 continuous minutes; order refill fluid below. Automatic shut-off after four minutes preserves battery life.
What’s included:
flow tube, rechargeable battery, battery charger/power supply, one fog refill can, and carrying case.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Fog Generator
Dimensions
FlowMarker:
8 1/4"W x 5 1/4"H x 3 1/2"D
Flow tube:
11 1/4"L x 0.79" dia
Brand
Degree Controls
Power
12V rechargeable Pb battery
Accessories
Qty
/
Price
Availability
Fog refill, 2.5 oz bottle
EW-10322-38
at
$141.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title