United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-26072-50
Fluke
®
725 Process Calibrator
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-26072-50
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-82
NIST-Traceable Calibration; Handheld Precision Process Calibrator
$170.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Calibration
>
Calibration Equipment
>
Fluke725 Multifunction Process Calibrator
Share
|
Fluke725 Process Calibrator
EW-26072-50
$2,499.95 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Measure and source functions allow you to test and calibrate almost anything!
Easy to read measure/source screen allows you view input and output simultaneously
Perform rapid linearity tests with auto step and auto ramp features
Rugged, streamlined design is ideal for field conditions
Easy to read measure/source screen allows you view input and output simultaneously
Perform rapid linearity tests with auto step and auto ramp features
Rugged, streamlined design is ideal for field conditions
Easy to read measure/source screen allows you view input and output simultaneously
Perform rapid linearity tests with auto step and auto ramp features
Rugged, streamlined design is ideal for field conditions
Use this multifunction process calibrator to measure volts, mA, RTD’s, thermocouples, frequency, and ohms to test sensors and transmitters. The dual channel functionality of this unit allows you to source/simulate volts, mA, thermocouples, RTD’s, frequency, ohms, and pressure to easily calibrate transmitters. Use this process calibrator to support flowmeter testing with the frequency and counts per minute functions provided. The frequently used function provides memory of test setups for future use and can store up to eight setups on any sourced function. Power your transmitters during test using the loop supply with simultaneous mA measurement.
This versatile process calibrator will measure/source pressure using any of the 29 Fluke 700Pxx pressure modules and has the ability to source mA with simultaneous pressure measurement to conduct valve and I/P tests. The rugged design is ideal for harsh field conditions and the simple pushbutton user interface is backlit for working in poor lighting conditions.
Available for sale in the United States and Canada only.
Specifications
Product Type
Process Calibrators/Meters
Resolution
0.01 mV 0.01 V (Source) 0.01 V (measure) 0.001 (mA) 0.01 to 0.1 ohm (Resistance) 2.0 to 1,000.0 CPM = 0.1 CPM (Frequency) 1 to 1100 Hz = 1 Hz (Frequency) 1.0 to 10.0 kHz = 0.1 kHz (Frequency) 0.1°C/F (Thermocouple - J, K, T, E, L, N, U) 1
Accuracy
±0.02% of rdg + LSD (Voltage) ±0.02% of rdg + LSD (mA) ±0.10 to 1.0 ohm (Resistance) ±0.05% (2.0 to 1,000.0 CPM) Frequency ±0.05% (1 to 100 Hz) Frequency ±0.25% (1.0 to 10.0 kHz) Frequency ±10% (Loop Supply) To 0.7<
Dimensions
3.75 in W x 7.875 in H x 1.875 in D
Power
Four AA batteries
Accessories
Qty
/
Price
Availability
Hard Case w/ Insulation
EW-20007-96
at
$59.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Hard Meter Case
EW-20008-00
at
$59.95 USD/EACH
In stock.
Add to Cart
Soft Meter Case
EW-20008-04
at
$69.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Fluke ToolPak Magnetic Meter Hanging Kit
EW-20008-56
at
$39.95 USD/EACH
In stock.
Add to Cart
Fluke TL220 Industrial Test Lead Set
EW-26002-30
at
$69.95 USD/SET
In stock.
Add to Cart
Fluke TL75 Right-Angle Test Lead Set
EW-26002-75
at
$20.95 USD/SET of 1
In stock.
Add to Cart
Carrying case; Fluke model C125
EW-26003-12
at
$39.95 USD/EACH
In stock.
Add to Cart
Leather Meter Carrying Case
EW-26003-60
at
$64.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Handheld Precision Process Calibrator
EW-17101-82
at $170.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
FLUKE 725 User Manual.pdf
Virtual Catalog (PDFs)
13-14 Catalog
More info
FLUKE 725 Calibration Manual.pdf
FLUKE 725 Safety Manual.pdf
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title