United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 9
of
12 >
Representative photo only
Fluke 190 202 Oscilloscope Handheld 2 Channel 200 MHz color
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-20041-34
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17100-33
NIST-Traceable Calibration; Oscilloscopes, up to 300MHz
$140.00 USD/EACH/EACH
Usually ships in 16 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Electrical
>
Oscilloscopes
>
Handheld Oscilloscopes
>
Fluke 190 Series II ScopeMeters
Share
|
Fluke 190-202 Oscilloscope, Handheld, 2-Channel, 200 MHz, color
EW-20041-34
$3,799.95 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
The first high-performance two and four-channel scopes built for harsh industrial environments
Rugged, IP 51 rating, dust- and drip-proof
Connect-and-View™ triggering for intelligent, automatic triggering on fast, slow, and even complex signals
Automatic capture and replay of 100 screens
30,000 points or more per input channel using ScopeRecord™ roll mode
TrendPlot™ Paperless recorder with deep memory for long-term automatic measurements
Frequency Spectrum using FFT-analysis
5,000 count DMM (2-channel models only)
These portable oscilloscopes combine rugged portability with the performance of a bench scope, taking you from troubleshooting microelectronics all the way into power electronic applications. Designed for plant maintenance engineers there is now a four-channel scope for the harsh world of industrial electronics that allow you to easily measure input, output, and feedback signals simultaneously to diagnose industrial electronics. Additionally, units allow you to safely troubleshoot three-phase systems like variable speed drives, as well as UPS or back-up generators. See more, fix more, and go where you’ve never gone before with a scope.
Four channel models additional features include four independent isolated inputs, up to 1000 V,
high-speed sampling - up to 2.5 GS/sec, deep memory: 10,000 points per trace waveform capture, up to 7 hours of battery operation, isolated USB host port for direct data storage to a USB memory device, USB device port for easy PC communication, and security lock down capabilities.
Available for sale in the US and Canada only
What's Included:
BC190 power adapter, BP291 double-capacity Li-ion battery pack (4-channel models), BP290 double capacity Li-ion battery pack (2-channel models), VPS410-R red probe set, VPS410-B blue probe set, VPS410-G gray probe set, VPS410-V green probe set, USB cable with mini B connector, hanging strap, side strap, CD user manual with FlukeView ScopeMeter software demo version, and calibration document supplied by the manufacturer.
Specifications
Product Type
Handheld Oscilloscopes
Bandwidth (MHz)
200
Input channels
2
Record length
27,500 pts per input
Display
Full-color high resolution backlit LCD (153 mm), 320 x 240 pixels
Power
Li-ion battery; 4-channel models (5200 mAh), 2-channel models (3600 mAh)
Inputs
2
Sample rate (Gs/s)
2.5
Channels
2
CE Compliance
Yes
Model
190-202
Make
Fluke
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Oscilloscopes, up to 300MHz
EW-17100-33
at $140.00 USD/EACH
Usually ships in 16 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
NIST method improves timing in oscilloscopes
Verifying hydraulic control system operation with Fluke ScopeMeter
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
ABCs of DMMs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title