dummy
SI-26071-60
- No need to carry two meterschoose a multimeter and current loop calibrator in one!
- Simulate up to 20 mA with current and percent of scale digital readouts
- Adjust current output in manual or automatic steps (25%) or set automatic slow/fast ramping
- Measures true-rms AC voltage and current
- Modes include continuity, relative, diode test and minimum/maximum/average recording
- Splash-proof and dust-proof housing protects meter in harsh conditions.
Available for sale in the United States and Canada only.
What's Included:
general purpose test leads (Fluke® model no. TL75), alligator clips (Fluke® model no. AC70A), protective holster (Fluke® model no. C81Y), and one 9 V battery
| Product Type |
Multimeter |
| AC voltage accuracy |
±0.7% of rdg + 2 digits |
| AC current accuracy |
±1% of rdg + 2 digits |
| DC current accuracy |
0 to 30 mA : ±0.05% ±0.2% of rdg + 2 digits |
| DC current range |
0 to 1.000A |
| DC voltage accuracy |
±0.1% of rdg + 1 digit |
| DC voltage range |
400.0 mV to 1000 V |
| Resistance range |
400.0 ohm to 40.00 M ohm |
| Frequency range |
199.99 Hz to 199.99 kHz |
| AC voltage range |
400.0 mV to 1000 V |
| Power |
9 volt battery |
| AC current range |
0 to 1.000 A |
| Dimensions |
3.41 in W x 7.35 in H x 1.25 in D |
| Make |
PROCESS TOOLS |
| Model |
FLUKE-787 |
| Manufacturer number |
FLUKE-787 |
| Brand |
Fluke |
at $25.60 USD/PACKAGE of 4
In stock.
at $69.95 USD/EACH
Usually ships in 14 days.
at $39.95 USD/EACH
In stock.
at $37.45 USD/SET
In stock.
at $32.45 USD/SET
In stock.
at $24.95 USD/SET
In stock.
at $69.95 USD/SET
In stock.
at $134.95 USD/EACH
In stock.
at $16.95 USD/SET
In stock.
at $49.95 USD/EACH
Usually ships in 14 days.
at $34.95 USD/EACH
In stock.
at $64.95 USD/EACH
Usually ships in 14 days.
at $129.95 USD/EACH
Usually ships in 14 days.
at $224.95 USD/EACH
In stock.
at $299.95 USD/EACH
In stock.
at $349.95 USD/EACH
In stock.
at $399.95 USD/EACH
In stock.
at $499.95 USD/EACH
In stock.
at $170.00 USD/EACH
Usually ships in 8 days.
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection