United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-26072-10
Fluke
®
789 Process Meter
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-26072-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-82
NIST-Traceable Calibration; Handheld Precision Process Calibrator
$170.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Calibration
>
Calibration Equipment
>
Fluke789 Process Meter
Share
|
Fluke 789 Process Meter
EW-26072-10
$919.95 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Unit provides simultaneous mA and % of scale readout on mA output
Meters feature 1000 V overload protection on Voltage, ohms, and frequency
Simulate up to 20 mA with current and percent of scale digital readouts
24 V loop power supply
mA source drive to 1200 ohms
HART® mode setting with loop power (adds 250 ohm resistor)
This combination multimeter and calibrator features an extra large display and built-in 250-ohm resistor, eliminating the need to carry a separate resistor. Infrared I/O serial port allows data to be exported to a PC. Diode and continuity testing has beeper signal.
Simultaneously displays mA and percent of scale. A 24 V loop power supply is provided. Unit features a large, dual display, and enhanced backlighting with two brightness settings.
Available for sale in the United States and Canada only.
What's Included:
Fluke TL71 test lead set, Fluke AC72 alligator clips, (4) AA alkaline batteries (installed), user’s manual and quick reference guide.
Specifications
Product Type
Process Calibrators/Meters
Range
VDC: 400.0 mV, 4.000 V, 40.00 V, 400.0 V, 1000 V
VAC (true-rms): 400.0 mV, 4.000 V, 40.00 V, 400.0 V, 1000 V
mA DC: 30.000 mA
ADC: 1.000 A (0.440 A continuous
AAC: 1.000 A (0.440 A continuous
Resistance: 400.0 Ohms, 4.000 k, 40.
Display
5-digit LCD
Operating temperature
-4 to 131°F (-20 to 55°C)
Accuracy
VDC: 0.1% + 1
VAC: 0.7% + 2
mA DC: 0.05% + 2
ADC: 0.2% + 2
AAC: 1% + 2
Frequency: 0.005% + 1
Diode test: 2% + 1
Dimensions
8 in L x 4 in W x 2 in H
Power
four AA batteries
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Fluke I1010 : Current Probe AC/DC Clamp-On
EW-20003-40
at
$374.95 USD/EACH
Usually ships in 14 days.
Add to Cart
AC Current Clamp
EW-20003-45
at
$139.95 USD/EACH
In stock.
Add to Cart
Hard Case w/ Insulation
EW-20007-96
at
$59.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Hard Meter Case
EW-20008-00
at
$59.95 USD/EACH
In stock.
Add to Cart
Soft Meter Case
EW-20008-04
at
$69.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Fluke ToolPak Magnetic Meter Hanging Kit
EW-20008-56
at
$39.95 USD/EACH
In stock.
Add to Cart
Fluke TL220 Industrial Test Lead Set
EW-26002-30
at
$69.95 USD/SET
In stock.
Add to Cart
Universal hard carrying case (Fluke model no. C100); polypropylene
EW-26003-10
at
$49.95 USD/EACH
Usually ships in 14 days.
Add to Cart
Carrying case, soft
EW-26003-40
at
$34.95 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Handheld Precision Process Calibrator
EW-17101-82
at $170.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title