dummy
EW-26087-02
- Diagnostic functions provide more troubleshooting power
- Unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment
- True RMS AC voltage and current for accurate measurements on nonlinear signals
- Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
- Measure up to 1000 VAC and DC
- Measure up to 10 A (20 A for 30 seconds)
- 10,000 µF capacitance range
- Frequency to 200 kHz and % duty cycle
- Peak capture to record transients as fast as 250 µs
- Large display digits and 2-level bright white backlight for increased visibility
- Withstands hazardous 8000 volt spikes caused by load switching and faults on industrial circuits and complies with second edition ANSI and IEC electrical safety standards
- Improved sleep mode for long battery life
- Relative mode to remove test lead resistance from low ohms measurements
- Auto and manual ranging for maximum flexibility
- Optional magnetic hanger (TPAK) for easy setup and viewing while freeing your hands for other tasks
Available for sale in the United States and Canada only
The Fluke 87V has improved measurement functions, troubleshooting features, resolution and accuracy to solve more problems on motor drives, in plant automation, power distribution, and electromechanical equipment. The 87V provides more problem-solving power, safety, convenience and impact protection.
The 87V is CAT III 1000 V and CAT IV 600 V rated and can withstand 8000 volt spikes caused by load switching on industrial circuits. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically and graphically with analog bar graph on the two-level, backlit LCD. Min/Max and average readings are recorded to capture variations. Meter features True-RMS measurements with the unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment. Unit also features a built-in temperature probe for temperature measurement capability.
What's Included:
TL75 test leads, AC72 alligator clips, temperature probe, CD-ROM (user's manual and technical notes), operator's guide, holster, and 9 V battery (installed).
| Product Type |
Multimeter |
| AC voltage accuracy |
±0.7%+2 True RMS |
| AC current accuracy |
±1.0%+2 True RMS |
| AC Max (Amps) |
10A (20 A for 30 seconds maximum) |
| AC Max (Voltage) |
1000 |
| DC current accuracy |
±0.2%+2 |
| DC current range |
10A (20 A for 30 seconds maximum) |
| DC voltage accuracy |
±0.05%+1 |
| DC voltage range |
1000 V |
| Capacitance accuracy |
±1.0%+2 |
| Current (AC/DC) range |
10 A |
| Frequency accuracy |
±0.005%+1 |
| Temp accuracy |
1.0% |
| Conductance |
50nS |
| Capacitance |
9,999µF |
| Count |
20000 (hi res mode) |
| Frequency |
199.99 kHz |
| Resistance |
50M ohm |
| Digits |
4.5 |
| Resistivity accuracy |
±0.2%+1 |
| Safety rating |
CAT III 1000V, CAT III 600V |
| Temperature |
-328 to 1994°F (-200 to 1090°C) |
| Make |
DMMS |
| Model |
FLUKE-87V |
| Manufacturer number |
FLUKE-87-5 |
| Brand |
Fluke |
at $25.60 USD/PACKAGE of 4
In stock.
at $139.95 USD/EACH
In stock.
at $99.95 USD/EACH
Usually ships in 14 days.
at $59.95 USD/EACH
Usually ships in 14 days.
at $59.95 USD/EACH
In stock.
at $69.95 USD/EACH
Usually ships in 14 days.
at $139.95 USD/EACH
In stock.
at $39.95 USD/EACH
In stock.
at $179.95 USD/EACH
Usually ships in 14 days.
at $224.95 USD/EACH
In stock.
at $39.95 USD/EACH
In stock.
at $34.95 USD/EACH
In stock.
at $23.95 USD/EACH
In stock.
at $75.00 USD/EACH
Usually ships in 6 days.
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334