United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
SI-37851-20
Fluke 971 Thermohygrometer
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-37851-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-28
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Humidity Products
>
Humidity Meters / Thermohygrometers Meters
>
Standard Thermohygrometers
>
Fluke 971 Thermohygrometer
Share
|
Fluke 971 Thermohygrometer
SI-37851-20
$274.95 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ergonomically designed meter with datalogger
Calculates dew point and wet bulb
Use as part of regular HVAC maintenance
The Fluke 971 meter is built for field use with an impact resistant housing, rugged holster, convenient belt clip, and bright backlit display. Includes degF/degC selection, Min/Max reading, hold feature, averaging function, and 99 record datalogging capacity. A large LCD simultaneously displays RH and temperature readings, along with calculating dew point and wet bulb.
Available for sale in the United States and Canada only.
What's Included:
four AAA batteries
Specifications
Product Type
Thermohygrometer
RH range
5 to 95%
RH accuracy
±2.5%
RH resolution
0.1%
Max temperature (º C)
60
Temp accuracy
±1.0°F @ 32 to 113°F, ±2.0°F for remaining range (±0.5°C 0 to 45°C, ±1.0°C for remaining range)
Temp resolution
0.2°F (0.1°C)
Response time
0.5 seconds
Display
RH: 3-digit LCD,Temperature: 3-digit LCD
Max temperature (º F)
140
Min temperature (º F)
-4
Min temperature (º C)
-20
Temp range
-4 to 140°F (-20 to 60°C)
RH range (Max)
95%
RH range (min)
5%
Dewpoint
Yes
Data Logging
Yes
Wet bulb
Yes
Dimensions
2.5 in W x 7.5 in H x 1.25 in D
Power
4 AAA battery (included)
Brand
Fluke
Manufacturer number
FLUKE-971
Model
FLUKE-971
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AAA, 12/pack
EW-09376-00
at
$21.25 USD/PACKAGE of 12
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
EW-17030-28
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Humidity Expressed as Dewpoint Temperature
Temperature FAQs
Temperature Instrument Ranges
Show All...
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title