dummy
EW-36302-02
Usually ships in 9 days.
Qty:
- Simple and safe operation for all flame related applications in the laboratory
- Ideal for use in all industrial, laboratory, and cleanroom settings
- Modern low-profile body design reduces air flow disturbances and is very easy to clean
- Robust, all stainless steel construction and fireproof controls
With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.Fuego SCS Basic Burner can be operated with a foot pedal or button function. Three programs are available: a
flexible start/stop function with 60-minute timer and the conventional foot-pedal operation for short flame sterilization. Additionally the flame can be started and stopped by pushing the function knob.(Foot pedal included.)
What's Included:
global power supply adapter (100 to 240 VAC, 50/60 Hz), tilt mechanism,
holding device for 3 inoculation loop holder and nozzles for natural gas, propane, and butane
| Product Type |
Burner |
| Gas type |
Natural Gas, Propane/Butane |
| Temperature |
1350°C on liquid gas, 1300°C on natural gas |
| Inlet tube OD |
1/4" |
| Dimensions |
103 mm W x 49 mm H x 130 mm D |
| Brand |
Fuego |
at $1,558.00 USD/EACH
Usually ships in 9 days.
at $46.00 USD/EACH
Usually ships in 7 days.
at $178.00 USD/EACH
Call 1-888-358-4705 for availability.
at $59.00 USD/EACH
Usually ships in 7 days.
at $29.25 USD/EACH
Usually ships in 10 days.
at $54.00 USD/EACH
Usually ships in 11 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334