Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Fuego SCS Pro -- with Infrared and rechargeable battery

EW-36302-06
$850.00 USD/EACH

Usually ships in 7 days.
Qty

  • Simple and safe operation for all flame related applications in the laboratory
  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.Fuego SCS Pro Burner offers all the features of the above models plus adds maximum flexibility through the state-of-the-art rechargeable battery technology. Unit includes an integrated fast charge function and two rechargeable batteries ( NiMH 2.4V, Mignon (AA)) for up to 9 hours of continuous use.

What's Included:
global power supply adapter (100 to 240 VAC, 50/60 Hz), tilt mechanism,
holding device for 3 inoculation loop holder and nozzles for natural gas, propane, and butane
Product Type Burner
Gas type Natural Gas, Propane/Butane
Temperature 1350°C on liquid gas, 1300°C on natural gas
Inlet tube OD 1/4"
Dimensions 103 mm W x 49 mm H x 130 mm D
Brand Fuego

Accessories
Qty/Price
Availability

at $1,558.00 USD/EACH

Usually ships in 9 days.

at $46.00 USD/EACH

Usually ships in 7 days.
Gas-Safety Adapter
EW-36302-11

at $178.00 USD/EACH

Call 1-888-358-4705 for availability.
Spray Protector
EW-36302-12

at $59.00 USD/EACH

Usually ships in 7 days.

at $29.25 USD/EACH

Usually ships in 10 days.

at $54.00 USD/EACH

Usually ships in 11 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title